Anti-Clostridium difficile Toxin A Mouse Monoclonal Antibody [clone: PCG4.1]
Artikelnr. ABNOMAB4039
Lieferant: Abnova
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Clostridium difficile Toxin A
- Klonalität:Monoklonal
- Klon:PCG4.1
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Isotyp:IgG2a kappa
- Reaktivität:Bacteria
- Kreuzadsorption:No
- Format:Liquid
- Lagerungspuffer:10 mM sodium phosphate buffer, 150 mM NaCl, pH 7,4 (0,05% sodium azide)
- Lagertemperatur:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Clostridium difficile Toxin A.
- VE:1 mg
Spezifikationen
Über diesen Artikel
Mouse monoclonal antibody raised against Clostridium difficile Toxin A.
Type: Primary
Antigen: Clostridium difficile Toxin A
Clonality: Monoclonal
Clone: PCG4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Bacteria