Order Entry
Austria
ContactUsLinkComponent
Anti-immunodeficiency virus type 1 Vif Mouse Monoclonal Antibody [clone: 319]
Catalog # ABNOMAB2431
Supplier:  Abnova
Anti-immunodeficiency virus type 1 Vif Mouse Monoclonal Antibody [clone: 319]
Catalog # ABNOMAB2431
Supplier:  Abnova

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    immunodeficiency virus type 1 Vif
  • Klonalität:
    Monoklonal
  • Klon:
    319
  • Konjugation:
    Unconjugated
  • Wirt:
    Mouse
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG1 kappa
  • Reaktivität:
    Virus
  • Western blot:
    Yes
  • Kreuzadsorption:
    No
  • Format:
    Liquid
  • Lagerungspuffer:
    PBS (0,1% sodium azide)
  • Lagertemperatur:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant His6 fusion protein corresponding to human immunodeficiency virus type 1 Vif.
  • Größe:
    100 μl
  • VE:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 Vif.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: immunodeficiency virus type 1 Vif
Clonality: Monoclonal
Clone: 319
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Virus