Anti-MAP3K5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13429
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Mitogen-activated protein kinase Kinase Kinase 5
- Antigen-Symbol:MAP3K5
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:Human,Mouse
- Western blot:Yes
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:4217
- Antigen-Synonyme:ASK1, ASK1, MEKK5, MAPKKK5, ASK1, Mitogen-activated protein kinase kinase kinase 5, Apoptosis signal-regulating kinase 1, ASK-1
- Lagerungspuffer:PBS (0,02% sodium azide)
- Lagertemperatur:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 16 amino acids of human MAP3K5.
- Größe:100 μg
- VE:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MAP3K5.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse