Anti-immunodeficiency virus type 1 P17 Gag Mouse Monoclonal Antibody [clone: 17-1]
Artikelnr. ABNOMAB2434
Lieferant: Abnova
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:immunodeficiency virus type 1 P17 Gag
- Klonalität:Monoklonal
- Klon:17-1
- Konjugation:Unconjugated
- Durchflusszytometrie:Yes
- Wirt:Mouse
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Isotyp:IgG2b kappa
- Western blot:Yes
- Kreuzadsorption:No
- Format:Liquid
- Lagerungspuffer:PBS (0,1% sodium azide)
- Lagertemperatur:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Human immunodeficiency virus P17 Gag protein.
- Größe:100 μl
- VE:100 µg
Spezifikationen
Über diesen Artikel
Mouse monoclonal antibody raised against human immunodeficiency virus P17 Gag protein.
Type: Primary
Antigen: immunodeficiency virus type 1 P17 Gag
Clonality: Monoclonal
Clone: 17-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: