Order Entry
Austria
ContactUsLinkComponent
 
Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-85]
  ABNOMAB4568
 :  Abnova
Anti-CD44 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-85]
  ABNOMAB4568
 :  Abnova

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CD44 molecule (Indian blood group)
  • Antigen-Symbol:
    CD44
  • Klonalität:
    Monoklonal
  • Klon:
    MEM-85
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Mouse
  • Isotyp:
    IgG2b
  • Reaktivität:
    Human
  • Kreuzadsorption:
    No
  • Format:
    Liquid
  • Gen-ID:
    960
  • Antigen-Synonyme:
    MDU2|HUTCH-I|LHR|MC56|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
  • Lagerungspuffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Lagertemperatur:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD44 from leukocytes of patient suffering from LGL-type leukaemia.
  • Größe:
    100 Reactions
  • VE:
    100 µl

 

 

Mouse monoclonal antibody raised against native CD44.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MEM-85
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human