Anti-TNFRSF10A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DR-4-02]
Catalog # ABNOMAB5077
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Tumor Necrosis Factor Receptor Superfamily, Member 10A
- Antigen-Symbol:TNFRSF10A
- Klonalität:Monoklonal
- Klon:DR-4-02
- Konjugation:PE (Phycoerythrin)
- Durchflusszytometrie:Yes
- Wirt:Mouse
- Isotyp:IgG1
- Reaktivität:Human
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:8797
- Antigen-Synonyme:TRAILR1|DR4|APO2|CD261|TRAILR-1
- Lagerungspuffer:PBS (0,2% BSA, 15 mM sodium azide)
- Lagertemperatur:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant Fc fusion protein corresponding to extracellular region of TNFRSF10A.
- Größe:100 μg
- VE:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant TNFRSF10A.
Recommended Dilutions: Flow Cytometry: 5 ?g/ml
Type: Primary
Antigen: TNFRSF10A
Clonality: Monoclonal
Clone: DR-4-02
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human