Anti-METTL7B Rabbit Polyclonal Antibody
ABNOPAB16592
: Abnova
- Antikörper-Typ:Primär
- Antigen-Name:Methyltransferase like 7B
- Antigen-Symbol:METTL7B
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Reaktivität:Human,Mouse
- Western blot:Yes
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:196410
- Antigen-Synonyme:ALDI
- Lagerungspuffer:PBS (0,02% sodium azide)
- Lagertemperatur:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus 13 amino acids of human METTL7B.
- Größe:100 μg
- VE:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of METTL7B.
Recommended Dilutions: Western Blot: 2-4 ?g/ml
Type: Primary
Antigen: METTL7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse