Anti-TP53INP2 Rabbit Polyclonal Antibody
Artikelnr. ABNOPAB17410
Lieferant: Abnova
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:Tumor Protein 53-inducible Nuclear Protein 2
- Antigen-Symbol:TP53INP2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Reaktivität:Human,Rat,Mouse
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:58476
- Antigen-Synonyme:PINH|DOR|C20orf110
- Lagerungspuffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Lagertemperatur:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human TP53INP2.
- Größe:100 μg
- VE:100 µg
Spezifikationen
Über diesen Artikel
Rabbit polyclonal antibody raised against synthetic peptide of TP53INP2.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Immunofluorescence: 1:500-1:1000; ELISA: 1:20000
Type: Primary
Antigen: TP53INP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat