Anti-SYN1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9658
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Synapsin 1
- Antigen-Symbol:SYN1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:Zebrafish,Human,Bovine,Xenopus laevis,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:24949
- Antigen-Synonyme:SYN1a|Synapsin-1|Synapsin I|SYN1|Brain protein 4.1|SYNI|SYN1b
- Modification:phospho-Ser603
- Lagerungspuffer:10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
- Lagertemperatur:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S603 of rat Syn1.
- Reinigung:Affinity purification
- Größe:100 μl
- VE:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Syn1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Clawed frog, Human, Mouse, Rat, Zebrafish