Colorimetric test kit, chloride, TESTOVAL®
HEYL410527
:
- VE:1
- Bezeichnung:Chloride, colour comparison kit
- Seq:01
- Messbereich:1-5-10-25-50-100 mg/l
Visual colour test method using a comparator disc.
Visual colour test method using a comparator disc.