Digital camera, Axiocam MRc
Artikelnr. ZEIS4265089901000
Lieferant: ZEISS
Spezifikationen
- Modell:AxioCam MRc
- Auflösung:1,4 MP
- Sensortyp:CCD
- Sensorgröße:2/3"
- Bezeichnung:Digital camera
- Pixelgröße:6,45×6,45 µm
- Pixel (B×H):1388×1040
- Betriebstemperatur:5...35 °C
- VE:1
Spezifikationen
Über diesen Artikel
High resolution camera made of blue anodized aluminum with cooling fins.
- One stage Peltier cooling
- Readout of sensor sub-regions (ROI)
- FireWire / IEEE1394 including cable and IR barrier filter BG40
- TTL control for controlling of external shutters
- Mid-range colour driver