Anti-PRDX3 Rabbit Polyclonal Antibody
Product Details & Documents
Rabbit IgG polyclonal antibody for Thioredoxin-dependent peroxide reductase, mitochondrial(PRDX3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
PRDX3(peroxiredoxin 3) also known as AOP-1, MER5, SP-22 or PRX3, is localized exclusively in mitochondria. The deduced 256-amino acid human AOP1 protein shares 86% amino acid sequence similarity with mouse Aop1, and significant similarity with both the human proliferation-associated gene A product and the mouse stress-induced peritoneal macrophage protein Msp23. The PRDX3 gene is mapped on 10q26.11. Expression of PRDX3 is induced by MYC and is reduced in c-myc -/- cells. Chromatin immunoprecipitation analysis spanning the entire PRDX3 genomic sequence revealed that MYC binds preferentially to a 930-bp region surrounding exon 1. Results using mitochondria-specific fluorescent probes demonstrated that PRDX3 is essential for maintaining mitochondrial mass and membrane potential in transformed rat and human cells. These data provided evidence that PRDX3 is a MYC target gene that is required to maintain normal mitochondrial function.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for Peroxiredoxin 3 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: PRDX6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Dokumente
PB9349
Technisches Datenblatt
Spezifikationen
- Größe:100 µg/vial
- Western blot:Yes
- Antikörper-Typ:Primär
- Form:Lyophilized
- Reinigung:Immunogen affinity purified.
- UniProtKB:P30048
- Reaktivität:Human|Rat|Mouse
- ImmunoChemie:Yes
- Cat. No.:BSBTPB9349
- Antigen-Name:peroxiredoxin 3
- Lagertemperatur:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Gen-ID:10935
- Immunogen:E.coli-derived human Peroxiredoxin 3 recombinant protein (Position: T110-Q256). Human Peroxiredoxin 3 shares 93% amino acid (aa) sequence identity with both mouse and rat Peroxiredoxin 3.
- Antigen-Synonyme:AOP1|AOP-1|SP-22|prx-III|PRO1748|MER5|HBC189
- Antigen-Symbol:PRDX3
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



