Centre well dish for IVF, Nunc™
Catalog # NUNC150260
Supplier: Thermo Fisher Scientific
Specifications
- Ø:55 mm
- Verpackt:Individually wrapped
- Bezeichnung:Centre well dish for IVF, vented, with lid
- Höhe:16 mm
- Environmentally Preferable:Umweltschonende Herstellung
- Umweltzertifizierung:NAR Certificate of Retirement
- VE:120
Specifications
About this item
Non treated surface, sterile (SAL 10⁻⁶), with lid. These IVF centre well dishes meet the exacting requirements of IVF testing, certification, and validation, providing consistently reliable, high quality results during embryo and oocyte washing.
- Perfect angle of reservoir well walls for locating and easy washing of samples
- True flat bottom for maintaining optimal temperature
- Easy grip design for increased safety
- Certificates of conformity available for every lot
Produced under ISO 13485 with full lot traceability
Certifications: Produced under ISO 13485 with full lot traceability.