Sterilisation indicator strips
Chemical indicator strip, for dry heat sterilisation, 160 - 180 °C
Anbieter: BAG Health Care
Springe zu:
Product Details & Documents
These indicator strips turns green to black colour.
- Visible conformation of item exposed to dry heat sterilisation process
- Monitors time and temperature
Certifications: Meets class I according to ISO 11140-1.
Spezifikationen
- Bezeichnung:Chemical indicator strip, for dry heat sterilisation, 160 - 180 °C