ContactUsLinkComponent
Pipettenspitzen, Mµlti Crystal
Artikelnr. SORE17000
Lieferant:  Sorenson Bioscience
undefined
Pipettenspitzen, Mµlti Crystal
Artikelnr. SORE17000
Lieferant:  Sorenson Bioscience
Spezifikationen
  • Verpackt:
    Lose (1000/Beutel)
  • Volumen:
    0,5 - 10 µl
  • Seq:
    01
  • VE:
    1.000
Spezifikationen
Über diesen Artikel
Virgin PP, non sterile, transparent.
  • Conical top and smooth narrow lumen for maximum sample recovery and accuracy
  • Manufactured from virgin materials and guaranteed free from pyrogens and heavy metals
  • Certified free from RNases, DNases, human DNA and PCR inhibitors
These 10 μl pipette tips are optimised to fit Eppendorf pipettes.