Oil test papers
Catalog # ROTHL832.1
Supplier: Roth Carl
CAS Number:
Specifications
- VE:100
- Bezeichnung:Oil Test Paper
Specifications
About this item
This test paper allows the rapid and reliable detection of oil contaminations of water and soil. The sensitivity largely depends on the nature of the respective hydrocarbon. For detection of oil in soil, press the paper firmly against the soil to be tested and rinse with clear water. To determine oil in water, move the paper back and forth a few times in the sample. In case of volatile hydrocarbons, the color reaction of the test paper has to be evaluated immediately.
The pale blue paper changes colour to dark blue upon contact with hydrocarbons, especially benzine, fuel oil, lubricating oil, and more.