Anti-Synuclein, pan Rabbit Polyclonal Antibody
USBIS9505-01A
- Antikörper-Typ:Primär
- Antigen-Symbol:Synuclein, pan
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:Human,Rat
- Western blot:Yes
- Form:lyophilized powder
- Lagerungspuffer:sterile water
- Lagertemperatur:Cool
- Konzentration:~1 mg/ml
- Transporttemperatur:Ice
- Größe:100 µl
- VE:100 µl
Type: Primary
Antigen: Synuclein, pan
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat