Anti-SUMO, Pan Rabbit Polyclonal Antibody (Biotin)
USBIS8026-01A-BIOT
- Antikörper-Typ:Primär
- Antigen-Symbol:SUMO, Pan
- Klonalität:Polyklonal
- Konjugation:Biotin
- ELISA:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Western blot:Yes
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:200 µl
- VE:200 µl
Type: Primary
Antigen: SUMO, Pan
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: