Anti-RAD9, pan Rabbit Polyclonal Antibody
USBIR0444-68
- Antikörper-Typ:Primär
- Antigen-Symbol:RAD9, pan
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Yeast
- Western blot:Yes
- Form:liquid
- Aminosäurezahl:1125 to 1139
- Lagerungspuffer:PBS, pH 7,2, 0,01% sodium azide.
- Lagertemperatur:Cool
- Konzentration:~0,7 mg/ml
- Transporttemperatur:Ice
- Reinigung:purified by immunoaffinity chromatography
- Größe:100 µg
- VE:100 µG
Recommended dilution: Immunoblot: 1:100-1:500; ELISA: 1:5000
Type: Primary
Antigen: RAD9, pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast