Anti-Histone H3, methyl, pan Rabbit polyclonal antibody
Catalog # USBIH5110-14N1
Supplier: US Biological
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Histone H3
- Antigen-Symbol:H3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Reaktivität:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:monomethyl-Lys9
- Lagerungspuffer:10mMsodium HEPES, pH 7,5, 150mM sodium chloride, 0,1mg/ml BSA and 50% glycerol.
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Reinigung:purified by immunoaffinity chromatography
- Größe:100 µl
- VE:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:25; Immunofluorescence: 1:500
Type: Primary
Antigen: Histone H3, methyl, pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat