Order Entry
Austria
ContactUsLinkComponent
 
Anti-F4/80, Pan Macrophages Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13A47]
  USBIF0001-51D
undefined
Anti-F4/80, Pan Macrophages Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13A47]
  USBIF0001-51D

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    F4/80, Pan Macrophages
  • Klonalität:
    Monoklonal
  • Klon:
    13A47
  • Konjugation:
    PE (Phycoerythrin)
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Rat
  • Isotyp:
    IgG2a
  • Reaktivität:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Lagerungspuffer:
    PBS, 1% BSA, 0,02% sodium azide.
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by Protein G chromatography
  • Größe:
    100 Tests
  • VE:
    100 Tests

 

 

Do not freeze!

Type: Primary
Antigen: F4/80, Pan Macrophages
Clonality: monoclonal
Clone: 13A47
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse