Anti-Endothelin, pan Rat monoclonal antibody [clone: 10C84]
USBIE3098-01R
- Antikörper-Typ:Primär
- Antigen-Name:Endothelin, pan
- Klonalität:Monoklonal
- Klon:10C84
- Konjugation:Unconjugated
- Wirt:Rat
- Isotyp:IgG2b
- Reaktivität:Human
- Form:liquid
- Lagerungspuffer:PBS, 0,09% sodium azide.
- Lagertemperatur:Cool
- Konzentration:~1 mg/ml
- Transporttemperatur:Ice
- Reinigung:purified by Protein G chromatography
- Größe:100 µg
- VE:100 µG
Type: Primary
Antigen: Endothelin, pan
Clonality: monoclonal
Clone: 10C84
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human