Order Entry
Austria
ContactUsLinkComponent
 
Anti-SPANX, pan Rabbit Polyclonal Antibody
  USBI168519
Anti-SPANX, pan Rabbit Polyclonal Antibody
  USBI168519

 

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    SPANX, pan
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Rabbit,
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Lagerungspuffer:
    0,02M potassium phosphate, 0,15M sodium chloride, pH 7,2, 0,01% sodium azide.
  • Lagertemperatur:
    Cool
  • Konzentration:
    ~1 mg/ml
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by immunoaffinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Recommended dilution: ELISA: 1:10000-1:50000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: SPANX, pan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rabbit