Order Entry
Austria
ContactUsLinkComponent
Anti-Lactate Dehydrogenase, pan Mouse Monoclonal Antibody [clone: 13F137]
Anti-Lactate Dehydrogenase, pan Mouse Monoclonal Antibody [clone: 13F137]
  USBI136679
Anti-Lactate Dehydrogenase, pan Mouse Monoclonal Antibody [clone: 13F137]
  USBI136679

 

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    Lactate Dehydrogenase, pan
  • Klonalität:
    Monoklonal
  • Klon:
    13F137
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Mouse
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Mouse
  • Form:
    liquid
  • Lagerungspuffer:
    0,1MPBS, pH 7,2, 0,09% sodium azide.
  • Lagertemperatur:
    Cool
  • Konzentration:
    ~0,5 mg/ml
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by Protein G chromatography
  • Größe:
    1 mg
  • VE:
    1 mg

 

 

Type: Primary
Antigen: Lactate Dehydrogenase, pan
Clonality: monoclonal
Clone: 13F137
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse