Anti-RPS4X Mouse Monoclonal Antibody [clone: 3E10]
Catalog # USBI132819
Supplier: US Biological
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Ribosomal Protein S4, X-linked
- Antigen-Symbol:RPS4X
- Klonalität:Monoklonal
- Klon:3E10
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Isotyp:IgM kappa
- Reaktivität:Human
- Format:ascites
- Form:liquid
- Aminosäurezahl:74 to 178
- Lagerungspuffer:ascites fluid.
- Sequence:GKVRTDITYPAGFMDVISIDKTGENFRLIYDTKGRFAVHRITPEEAKYKLCKVRKIFVGTKGIPHLVTHDARTIRYPDPLIKVNDTIQIDLETGKITDFIKFDT
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:200 µl
- VE:200 µl
Specifications
About this item
Type: Primary
Antigen: RPS4X (ribosomal protein S4, X-linked)
Clonality: monoclonal
Clone: 30000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Human