Order Entry
Austria
ContactUsLinkComponent
 
Anti-SERBP1 Mouse Monoclonal Antibody [clone: 1D2-2E9]
  USBI130833
Anti-SERBP1 Mouse Monoclonal Antibody [clone: 1D2-2E9]
  USBI130833

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    SERPINE1 mRNA binding protein 1
  • Antigen-Symbol:
    SERBP1
  • Klonalität:
    Monoklonal
  • Klon:
    1D2-2E9
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Mouse
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG1 kappa
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Form:
    liquid
  • Lagerungspuffer:
    PBS, pH 7,2.
  • Sequence:
    MPGHLQEGFGCVVTNRFDQLFDDESDPFEVLKAAENKKKEAGGGGVGGPGAKSAAQAAAQTNSNAAGKQLRKESQKDRKNPLPPSVGVVDKKEETQPPVALKKEGIRRVGRRPDQQLQGEGKIIDRRPERRPPRERRFEKPLEEKGEGGEFSVDRPIIDRPIRGRGGLGRGRGGRGRGMGRGDGFDSRGKREFDRHSGSDRSGLKHEDKRGGSGSHNWGTVKDELTESPKYIQKQISYNYSDLDQSNVTEETPEG
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by Protein A chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: SERBP1 (SERPINE1 mRNA binding protein 1)
Clonality: monoclonal
Clone: 1D2-2E9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human