Order Entry
Austria
ContactUsLinkComponent
Anti-GC Mouse Monoclonal Antibody [clone: 660CT8.6.6.2]
Catalog # USBI030988
Supplier:  US Biological
undefined
Anti-GC Mouse Monoclonal Antibody [clone: 660CT8.6.6.2]
Catalog # USBI030988
Supplier:  US Biological

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Group-specific component
  • Antigen-Symbol:
    GC
  • Klonalität:
    Monoklonal
  • Klon:
    660CT8.6.6.2
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Wirt:
    Mouse
  • Isotyp:
    IgM
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Epitop:
    internal
  • Format:
    ascites
  • Form:
    liquid
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Größe:
    100 µl
  • VE:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1600

Type: Primary
Antigen: GC (group-specific component (vitamin D binding protein))
Clonality: Monoclonal
Clone: 660CT8.6.6.2
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgM
Reactivity: Human