Anti-Neuronal Marker, Pan Rabbit Polyclonal Antibody (Biotin)
USBI029558
- Antikörper-Typ:Primär
- Antigen-Name:Neuronal Marker, Pan
- Klonalität:Polyklonal
- Konjugation:Biotin
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Reaktivität:Rat,Mouse
- Form:liquid
- Lagerungspuffer:PBS, 15mg/ml BSA, 0,05% sodium azide.
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Reinigung:purified
- Größe:100 µl
- VE:100 µl
Type: Primary
Antigen: Neuronal Marker, Pan
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat