Order Entry
Austria
ContactUsLinkComponent
Anti-pan PDK Mouse Monoclonal Antibody [clone: 3H5BE12]
Anti-pan PDK Mouse Monoclonal Antibody [clone: 3H5BE12]
  ABCAAB115321-100
 :  Abcam
undefined
Anti-pan PDK Mouse Monoclonal Antibody [clone: 3H5BE12]
  ABCAAB115321-100
 :  Abcam

 

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    PDK, pan
  • Klonalität:
    Monoklonal
  • Klon:
    3H5BE12
  • Konjugation:
    Unconjugated
  • Wirt:
    Mouse
  • Isotyp:
    IgG2b
  • Western blot:
    Yes
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Größe:
    100 µg
  • Packung:
    Vial
  • VE:
    100 µG

 

 

Mouse monoclonal [3H5BE12] to pan PDK.

 : All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: pan PDK
Clonality: Monoclonal
Clone: 3H5BE12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human; Rat; Mouse