Order Entry
Austria
ContactUsLinkComponent
Anti-pan CEACAM Mouse Monoclonal Antibody [clone: TET2]
Catalog # ABCAAB26283-50
Supplier:  Abcam
undefined
Anti-pan CEACAM Mouse Monoclonal Antibody [clone: TET2]
Catalog # ABCAAB26283-50
Supplier:  Abcam

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    CEACAM, pan
  • Klonalität:
    Monoklonal
  • Klon:
    TET2
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Mouse
  • ImmunoChemie:
    Yes
  • Immunpräzipitation:
    Yes
  • Isotyp:
    IgG2b
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Größe:
    50 µg
  • Packung:
    Vial
  • VE:
    50 µG

Specifications

About this item

Mouse monoclonal [TET2] to pan CEACAM.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: pan CEACAM
Clonality: Monoclonal
Clone: TET2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human