Anti-SLC12A2 Mouse Monoclonal Antibody
Artikelnr. BIRBORB95248-50
Lieferant: Biorbyt
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:solute carrier family 12 (sodium/potassium/chloride transporter), member 2
- Antigen-Symbol:Solute carrier family 12, member 2
- Klonalität:Monoklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- ImmunoChemie:Yes
- Isotyp:IgG2b kappa
- Reaktivität:Human
- Western blot:Yes
- Lagerungspuffer:PBS, pH 7,2
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Reinigung:purified by Protein A chromatography
- Größe:50 µg
- VE:50 µG
Spezifikationen
Über diesen Artikel
Immunochemistry: 5 ?g/ml
Type: Primary
Antigen: SLC12A2 (solute carrier family 12 (sodium/potassium/chloride transporter), member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human