Anti-HS1BP3 Rabbit Polyclonal Antibody
BIRBORB77868-100
: Biorbyt
- Antikörper-Typ:Primär
- Antigen-Name:HCLS1 binding protein 3
- Antigen-Symbol:HS1BP3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Immunogen:Recombinant protein
- Reinigung:purified by affinity chromatography
- Größe:100 µg
- VE:100 µG
ELISA: 1:20000-1:80000; Western Blot: 1:500-1:1000
Type: Primary
Antigen: HS1BP3 (HCLS1 binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human