Order Entry
Austria
ContactUsLinkComponent
 
Anti-ZNF329 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-ZNF329 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB126513-200
  • BIRBORB126513-100
  • Antigen-Symbol:
  • ZNF329
  • ZNF329
  • Antigen-Name:
  • zinc finger protein 329
  • zinc finger protein 329
  • Antikörper-Typ:
  • Primär
  • Primär
  • Klonalität:
  • Polyklonal
  • Polyklonal
  • Konjugation:
  • Unconjugated
  • Unconjugated
  • Reaktivität:
  • Human
  • Human
  • Wirt:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • Reinigung:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Konzentration:
  • 1 mg/ml
  • 1 mg/ml
  • Lagerungspuffer:
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Lagertemperatur:
  • Cool
  • Cool
  • Transporttemperatur:
  • Ice
  • Ice

 

 

Frequently Bought Together