Order Entry
Austria
ContactUsLinkComponent
Anti-TFF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-TFF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB110437-100
Supplier:  Biorbyt
Anti-TFF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB110437-100
Supplier:  Biorbyt

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Trefoil factor 2
  • Antigen-Symbol:
    TFF2
  • Klonalität:
    Polyklonal
  • Konjugation:
    HRP (Horseradish Peroxidase)
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Form:
    liquid
  • Lagerungspuffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Lagertemperatur:
    Cool
  • Transporttemperatur:
    Ice
  • Immunogen:
    Synthetic peptide
  • Reinigung:
    purified by affinity chromatography
  • Größe:
    100 µl
  • VE:
    100 µl

Specifications

About this item

Type: Primary
Antigen: TFF2 (trefoil factor 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: