Order Entry
Austria
ContactUsLinkComponent
Anti-MAPT Rabbit Polyclonal Antibody
Catalog # BIRBORB106033-100
Supplier:  Biorbyt
undefined
Anti-MAPT Rabbit Polyclonal Antibody
Catalog # BIRBORB106033-100
Supplier:  Biorbyt

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    microtubule-associated Protein tau
  • Antigen-Symbol:
    MAPT
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Modification:
    phospho-Ser404
  • Lagerungspuffer:
    Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Molecular weight:
    78 kDa
  • Lagertemperatur:
    Cool
  • Konzentration:
    20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
  • Transporttemperatur:
    Ice
  • Immunogen:
    A synthesized peptide derived from human Tau around the phosphorylation site of Serine 404
  • Reinigung:
    purified by immunoaffinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

Western Blot: 1:500-1:2000

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat