Anti-WNT10A Rabbit Polyclonal Antibody
Artikelnr. BIRBORB41669-100
Lieferant: Biorbyt
Spezifikationen
- Antikörper-Typ:Primär
- Antigen-Name:wingless-type MMTV integration site family, member 10A
- Antigen-Symbol:WNT10A
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Aminosäurezahl:47 to 241
- Lagerungspuffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Lagertemperatur:Cool
- Konzentration:1 mg/ml
- Transporttemperatur:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 47 and 241 of human WNT10A.
- Reinigung:purified by antigen affinity chromatography
- Größe:100 µg
- VE:100 µG
Spezifikationen
Über diesen Artikel
Western Blot: 1:500-1:3000
Type: Primary
Antigen: WNT10A (wingless-type MMTV integration site family, member 10A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human