Anti-HDL Rabbit Polyclonal Antibody
: Biorbyt
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HDL
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- BIRBORB5392-200
- BIRBORB5392-100
- Antigen-Symbol:
- HDL
- HDL
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Polyklonal
- Polyklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Human
- Human
- Wirt:
- Rabbit
- Rabbit
- Isotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- Immunogen:
- KLH conjugated synthetic peptide derived from human HDL.
- KLH conjugated synthetic peptide derived from human HDL.
- Form:
- liquid
- liquid
- Molecular weight:
- 31 kDa
- 31 kDa
- Reinigung:
- purified by affinity chromatography
- purified by affinity chromatography
- Konzentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Lagerungspuffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Lagertemperatur:
- Cool
- Cool
- Transporttemperatur:
- Ice
- Ice
Frequently Bought Together