Order Entry
Austria
ContactUsLinkComponent
Anti-HSP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB3703-100
Supplier:  Biorbyt
Anti-HSP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB3703-100
Supplier:  Biorbyt

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    HSP1
  • Klonalität:
    Polyklonal
  • Konjugation:
    FITC (Fluorescein Isothiocyanate)
  • Wirt:
    Rabbit
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Form:
    liquid
  • Lagerungspuffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Lagertemperatur:
    Cool
  • Konzentration:
    0,5 mg/ml
  • Transporttemperatur:
    Ice
  • Immunogen:
    Synthetic peptide
  • Reinigung:
    purified by Protein A chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: HSP1
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human