Anti-MARCH8 Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: MARCH8 (membrane-associated ring finger (C3HC4) 8, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- BIRBORB1445-100
- BIRBORB1445-200
- Antigen-Symbol:
- MARCH2
- MARCH3
- Antigen-Name:
- Membrane associated ring-CH-type finger 2
- Membrane associated ring-CH-type finger 3
- Antigen-Synonyme:
- RNF172|MARCH-II|HSPC240
- RNF173|MARCH-III
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Polyklonal
- Polyklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Wirt:
- Rabbit
- Rabbit
- Isotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- Immunogen:
- Synthetic peptide
- Synthetic peptide
- Form:
- liquid
- liquid
- Reinigung:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Konzentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Lagerungspuffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Lagertemperatur:
- Cool
- Cool
- Transporttemperatur:
- Ice
- Ice
Specifications
Frequently Bought Together