Anti-ZFP3 Mouse Polyclonal Antibody
Catalog # ABNOH00124961-B03
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Zinc Finger Protein 3
- Antigen-Symbol:ZFP3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Mouse
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:124961
- Antigen-Synonyme:ZNF752
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:50 µl
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: ZFP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human