Anti-HIPK2 Mouse Monoclonal Antibody [clone: 1F10]
ABNOH00028996-M03
: Abnova
- Antikörper-Typ:Primär
- Antigen-Name:HomeoDomain Interacting Protein kinase 2
- Antigen-Symbol:HIPK2
- Klonalität:Monoklonal
- Klon:1F10
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- ImmunoChemie:Yes
- Reaktivität:Human,Rat
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:28996
- Antigen-Synonyme:PRO0593|EC 2.7.11.1|HHIPK2
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µg
- VE:100 µg
Type: Primary
Antigen: HIPK2
Clonality: Monoclonal
Clone: 1F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat