Anti-RPS2 Mouse Monoclonal Antibody [clone: 3F5]
ABNOH00006187-M05
: Abnova
- Antikörper-Typ:Primär
- Antigen-Name:Ribosomal Protein S2
- Antigen-Symbol:RPS2
- Klonalität:Monoklonal
- Klon:3F5
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:6187
- Antigen-Synonyme:LLREP3|S2
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µg
- VE:100 µg
Type: Primary
Antigen: RPS2
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human