Anti-MPG Mouse Monoclonal Antibody [clone: 4B12]
Catalog # ABNOH00004350-M08
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:N-Methylpurine DNA Glycosylase
- Antigen-Symbol:MPG
- Klonalität:Monoklonal
- Klon:4B12
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- ImmunFluoreszenz:Yes
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:4350
- Antigen-Synonyme:MPG|3' end of the Mid1 gene|CRA36.1|localized 68 kb upstream the humanzeta globin gene on 16p|AAG|proliferation-inducing protein 16|Mid1|alkyladenine DNA glycosylase|MDG|PIG11|N-methylpurine-DNA glycosylase|anpg|proliferation-inducing protein 11|APNG|CRA36.1 (3-methyl-adenine DNA glycosylase)|PIG16
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µg
- VE:100 µg
Specifications
About this item
Type: Primary
Antigen: MPG
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human