Anti-GPC5 Mouse Monoclonal Antibody [clone: 1B8]
ABNOH00002262-M02
: Abnova
- Antikörper-Typ:Primär
- Antigen-Name:Glypican 5
- Antigen-Symbol:GPC5
- Klonalität:Monoklonal
- Klon:1B8
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Immunpräzipitation:Yes
- Reaktivität:Human
- Kreuzadsorption:No
- Gen-ID:2262
- Antigen-Synonyme:Glypican-5|GPC5
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µg
- VE:100 µg
Type: Primary
Antigen: GPC5
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human