Anti-MAFB Rabbit Polyclonal Antibody
Catalog # AVIVARP58149_P050
Supplier: Aviva Systems Biology
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B
- Antigen-Symbol:MAFB
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Kreuzadsorption:none
- Gen-ID:9935
- Antigen-Synonyme:KRML|MCTO
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:50 µG
Specifications
About this item
Type: Primary
Antigen: MAFB (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: