Anti-ANGPTL4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-5027R
Supplier: Bioss
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Angiopoietin-like 4
- Antigen-Symbol:ANGPTL4
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 µl
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: ANGPTL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat