Anti-CYP11B1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3898R
Supplier: Bioss
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Cytochrome P450, Family 11, Subfamily B, Polypeptide 1
- Antigen-Symbol:CYP11B1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:CPN1|P450C11|cytochrome P450|cytochrome P-450c11|cytochrome P450 11B1|subfamily XIB (steroid 11-beta-hydroxylase)|FHI|cytochrome p450 XIB1|polypeptide 1|cytochrome P450C11|mitochondrial|steroid 11-beta-monooxygenase|CYP11B|steroid 11-beta-hydroxylase|CYPXIB1
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: CYP11B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: