Anti-TLR2 Rabbit Polyclonal Antibody
BOSSBS-1019R
: Bioss
- Antikörper-Typ:Primär
- Antigen-Name:Toll-like Receptor 2
- Antigen-Symbol:TLR2
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:CD282|TIL4|Toll/interleukin-1 receptor-like protein 4
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:100 µl
Type: Primary
Antigen: TLR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine