Anti-IFNG Rabbit Polyclonal Antibody
Catalog # BOSSBS-0480R
Supplier: Bioss
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Interferon gamma
- Antigen-Symbol:IFNG
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat