Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog # BOSSBS-0222R
Supplier: Bioss
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Glutamate Ionotropic Receptor NMDA Type Subunit 2B
- Antigen-Symbol:GRIN2B
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Durchflusszytometrie:Yes
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Kreuzadsorption:none
- Antigen-Synonyme:hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B|glutamate ionotropic receptor NMDA type subunit 2B
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- VE:100 µl
Specifications
About this item
Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat